Skip to Content
Merck
All Photos(1)

Key Documents

CRM48641

Supelco

Naphthalene solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
7822574
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-30°C

SMILES string

c1ccc2ccccc2c1

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H

InChI key

UFWIBTONFRDIAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qianqian Shi et al.
Journal of environmental sciences (China), 25(1), 188-194 (2013-04-17)
A high-surface-area carbon (KC-1) was prepared from waste polystyrene-based ion exchange resin by KOH activation and used for naphthalene adsorption. The carbon exhibited a good hydrophobic nature with developed porous structure, favoring the adsorption of organic compounds. The Brunauer-Emmett-Teller surface
Fabrice Gritti et al.
Journal of chromatography. A, 1280, 35-50 (2013-02-07)
The achievable separation speed and resolution of columns packed with the new 4.6 μm Kinetex particles were characterized by their permeability and their plate heights, measured meticulously. Their specific permeabilities range between 1.81 and 1.95 × 10(-10)cm(2) and their external
Azusa Suzuki et al.
Bioorganic & medicinal chemistry letters, 23(3), 886-892 (2013-01-01)
We synthesized various C8-naphthylethynylated 2'-deoxyadenosine derivatives and investigated their photophysical properties. Among them, cyano- and N,N-dimethylamino-substituted 8-naphthylethynylated 2'-deoxyadenosine derivatives ((cn)A and (dn)A) showed strong fluorescence with high quantum yields and a remarkable solvatofuorochromicity. In particular, fluorescence of N,N-dimethylamino-substituted (2,6dn)A was
K Vijayavel et al.
Biomedical and environmental sciences : BES, 26(4), 295-302 (2013-03-29)
To investigate protective effect of Coleus aromaticus leaf extract against naphthalene induced hepatotoxicity in rats. Eighteen male rats were divided into three groups. Group I rats were treated as control. Group II rats were intraperitoneally administered with naphthalene (435 mg/kg
Dickson Y S Yan et al.
Environmental pollution (Barking, Essex : 1987), 178, 15-22 (2013-03-26)
The effectiveness and mechanisms of naphthalene and metal removal from artificially contaminated soil by FeEDTA/FeEDDS-activated persulfate were investigated through batch experiments. Using FeEDTA-activated persulfate, higher naphthalene removal from the soil at 7 h was achieved (89%), compared with FeEDDS-activated persulfate (75%).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service