Skip to Content
Merck
All Photos(1)

Key Documents

44637-U

Supelco

2-Methylnaphthalene solution

certified reference material, 1000 μg/mL in methanol

Synonym(s):

β-Methylnaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C11H10/c1-9-6-7-10-4-2-3-5-11(10)8-9/h2-8H,1H3

InChI key

QIMMUPPBPVKWKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radosław Swiercz et al.
International journal of occupational medicine and environmental health, 23(4), 385-389 (2011-02-11)
The aim of the study was to evaluate the toxicokinetics of 2-methylnaphtalene (2-MN) during and after inhalation exposure. Male Wistar rats were exposed to 2-MN vapours at nominal concentrations of 200 or 400 mg/m3 in the dynamic inhalation chamber for
Michael Safinowski et al.
Environmental microbiology, 8(2), 347-352 (2006-01-21)
The sulfate-reducing culture N47 can utilize naphthalene or 2-methylnaphthalene as the sole carbon source and electron donor. Here we show that the initial reaction in the naphthalene degradation pathway is a methylation to 2-methylnaphthalene which then undergoes the subsequent oxidation
Gro Harlaug Olsen et al.
Marine environmental research, 72(4), 179-187 (2011-09-13)
Reliable risk assessment approaches for Arctic environments are requested to manage potential impacts associated with increased activities in Arctic regions. We performed toxicity tests on Arctic and temperate species exposed to the narcotic acting oil component, 2-methyl naphthalene. The experimental
Drazenka Selesi et al.
Journal of bacteriology, 192(1), 295-306 (2009-10-27)
The highly enriched deltaproteobacterial culture N47 anaerobically oxidizes the polycyclic aromatic hydrocarbons naphthalene and 2-methylnaphthalene, with sulfate as the electron acceptor. Combined genome sequencing and liquid chromatography-tandem mass spectrometry-based shotgun proteome analyses were performed to identify genes and proteins involved
Young-Bum Kim et al.
Applied biochemistry and biotechnology, 159(1), 1-10 (2009-02-14)
Amphiphilic polyurethane (APU) nano-particle enhanced the mobilization of 2-methylnaphthalene (2-MNPT) in soil. Significant increase in the solubility of 2-MNPT was achieved. The molar solubilization ratio was 0.4 (mole 2-MNPT/mole APU). Simple precipitation of APU particle by 2 N CaCl(2) recovered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service