Skip to Content
Merck
All Photos(1)

Key Documents

13101AST

Supelco

Astec® CHIROBIOTIC® R Chiral (5 μm) HPLC Columns

L × I.D. 2 cm × 1 mm, HPLC Guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Astec® CHIROBIOTIC® R Chiral HPLC Guard Column, Column, 5 μm particle size, L × I.D. 2 cm × 1 mm

material

stainless steel column

Agency

suitable for USP L63

description

HPLC guard column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

2 cm × 1 mm

matrix

fully porous particle

matrix active group

ristocetin A glycopeptide phase

particle size

5 μm

pore size

100 Å

operating pH

3.5-6.8

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Henry et al.
Biomedical chromatography : BMC, 26(4), 425-428 (2011-08-16)
D-lactic acid in urine originates mainly from bacterial production in the intestinal tract. Increased D-lactate excretion as observed in patients affected by short bowel syndrome or necrotizing enterocolitis reflects D-lactic overproduction. Therefore, there is a need for a reliable and
P Dehouck et al.
Journal of chromatography. A, 1010(1), 63-74 (2003-09-25)
Erythromycin is a mixture of macrolide antibiotics produced by Saccharopolyspora erythreas during fermentation. A new method for the analysis of erythromycin by liquid chromatography has previously been developed. It makes use of an Astec C18 polymeric column. After validation in
B DeCruze et al.
Clinical oncology (Royal College of Radiologists (Great Britain)), 11(4), 252-254 (1999-09-04)
Poor prognosis (poorly differentiated and/or deep myometrial invasion) Stage I endometrial cancer can have a relapse rate as high as 50%. Traditionally, most clinical oncologists treat these patients with external beam radiotherapy after surgery but there is no evidence to
Mohie M K Sharaf El-Din et al.
Talanta, 189, 86-91 (2018-08-09)
In this study, the chromatographic conditions for separation and determination of L- and D-lactic acid enantiomers by copper ligand exchange chromatography have been examined and optimized statistically using a response surface methodology (RSM). The chromatographic variables: copper sulfate, acetic acid
G Bellucci et al.
Chirality, 6(3), 207-212 (1994-01-01)
The rabbit liver microsomal biotransformation of alpha-methylstyrene (1a), 2-methyl-1-hexene (1b), 2,4,4-trimethyl-1-pentene (1c), and 1,3,3-trimethyl-1-butene (1d) has been investigated with the aim at establishing the enantioface selection of the cytochrome P-450-promoted epoxidation of the double bond and the enantioselectivity of microsomal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service