Z722103
Pall Nanosep® centrifugal device with Omega membrane
MWCO 30 kDa
Synonym(s):
centifuge device, centrifugal devices, centrifugal filters
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
Recommended Products
material
polyethersulfone membrane
polypropylene housing
sterility
non-sterile
packaging
pack of 24
manufacturer/tradename
Pall OD030C33
tube size
~1.5 mL
working volume
0.5 mL
pore size
30 kDa MWCO
Looking for similar products? Visit Product Comparison Guide
General description
- Rapid processing of samples.
- Typical recoveries are greater than 90%.
- A wide range of MWCOs color-coded for easy identification.
- Constructed of low binding polypropylene.
- Ultrasonically welded seals prevent bypass or seal failure.
- Fits standard centrifuge rotors that accept 1.5 mL tubes.
Legal Information
Nanosep is a registered trademark of Pall Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service