Z618683
Eppendorf® UVette®
Starter set
Synonym(s):
biophotometer, cuvette uv, uv cuvette
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
Set containing 80 UVette + 1 universal adapter for 15 mm light center height; convertible to 8.5 mm
Starter set
manufacturer/tradename
Eppendorf® 4099 100.007
technique(s)
UV/Vis spectroscopy: suitable
photometry: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Two optical path lengths with a single rotation! Samples with a normal concentration can be measured using the 10 mm optical path length. For samples with a higher concentration, simply rotate the UVette by 90° and use the shorter 2 mm optical path length. 4 optical windows, crystal clear, UV and VIS transparent in the range 200-1600 nm. Funnel-shaped base prevents capillary effect, ensures sample in center of measuring area. The new UVette is ideal for use with the Eppendorf BioPhotometer. In other photometers or spectrometers the Uvette is inserted with the aid of an adapter. This adapter aligns the UVette to the corresponding height of the optical light path and also serves as an aperture for optimal light guidance and use of the small measuring volume. Individually packaged, UVettes are DNA, RNase, and protein-free, and are protected from scratches and dust.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
UVette is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service