Skip to Content
Merck
All Photos(1)

Key Documents

U8882

Sigma-Aldrich

Ubiquitin-Carrier Protein H5c human

≥95% (SDS-PAGE), recombinant, expressed in E. coli (GST-tagged), solution

Synonym(s):

UbcH5c

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in E. coli (GST-tagged)

Assay

≥95% (SDS-PAGE)

form

solution

mol wt

47 kDa

technique(s)

ligand binding assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UBE2D3(7323)

General description

Manufactured for Sigma by Boston Biochem., Inc.

Application

Ubiquitin-Carrier Protein H5c human may be used to study ubiquitin-proteasome system and its role in pathogenesis of human diseases.

Biochem/physiol Actions

Human homologs of the yeast UBC4/5 family and play many important regulatory roles in inflammation and cancer.
Mediates degradation of a myriad of short-lived regulatory proteins such as tumor suppressor p53.
Mediates the degradation of a myriad of short-lived regulatory proteins including the tumor suppressor p53 in the presence of E6/E6-AP or MDM2, the c-Fos transcription factor, and the signal-induced ubiquitination of IκBα, UBC5 is an essential gene in yeast.
Ubiquitin-Carrier Protein H5c human also referred to as UbcH5c is a recombinant protein that interacts with ZIPK and stimulate its ubiquitination and hence facilitates in regulating ZIPK accumulation in promyelocytic leukemia protein nuclear body (PML-NBs). [1]

Physical form

Solution in 50 mM HEPES, pH 7.6, 50 mM NaCl, 1 mM DTT in 10% glycerol.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norihiko Ohbayashi et al.
Biochemical and biophysical research communications, 372(4), 708-712 (2008-06-03)
Zipper-interacting protein kinase (ZIPK) is a widely expressed serine/threonine kinase that has been implicated in cell death and transcriptional regulation, but its mechanism of regulation remains unknown. In our previous study, we showed that leukemia inhibitory factor stimulated threonine-265 phosphorylation
H Gonen et al.
The Journal of biological chemistry, 274(21), 14823-14830 (1999-05-18)
The last step in the activation of the transcription factor NF-kappaB is signal-induced, ubiquitin- and proteasome-mediated degradation of the inhibitor IkappaBalpha. Although most of the components involved in the activation and degradation pathways have been identified, the ubiquitin carrier proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service