Skip to Content
Merck
All Photos(1)

Key Documents

SRP6538

Sigma-Aldrich

Lipocalin-2/LCN2 from rat

recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE)

Synonym(s):

24p3, MSFI, NGAL, Neutrophil Gelatinase-associated Lipocalin, Oncogene 24p3, Siderocalin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

rat

recombinant

expressed in HEK 293 cells

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

monomer ~250 kDa (Rat lipocalin-2 aa 1-198 is fused at the C-terminus to a FLAG0174-tag.)

packaging

pkg of 10 and 50 μg

impurities

<0.1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

Lipocalin-2 (LCN2), also known as oncogene 24p3 or neutrophil gelatinase-associated lipocalin (NGAL), MSFI, The binding of lipocalin-2 to bacterial siderophores is important in the innate immune response to bacterial infection. Upon encountering invading bacteria the toll-like receptors on immune cells stimulate the synthesis and secretion of lipocalin-2. Secreted lipocalin-2 then limits bacterial growth by sequestering iron-containing siderophores. Lipocalin-2 also functions as a growth factor. LCN2 is strongly upregulated during inflammation and is upregulated by interleukin 1 (but not TNF α) in humans. There are indications that some forms of acne could be caused due to the gene not being transcribed, and that Isotretinoin corrects this. NFAT3 (NFATc4) NFAT by blocking the expression of LCN2 inhibits breast carcinoma cell motility. Recent studies have revealed that NGAL plays an important role in the physiopathology of chronic myeloid leukaemia (CML) mediated by BCR-ABL.

Physical form

0.2 μm-filtered solution in PBS.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service