SRP5080
PTPRF (1275-1897), active, GST tagged human
recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
recombinant
expressed in E. coli
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
386-522 nmol/min·mg
mol wt
~93 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PTPRF(5792)
General description
PTPRF or LAR is a member of the protein tyrosine phosphatase family with an extracellular region, a single transmembrane region, and two tandem intracytoplasmic catalytic domains. PTPRF has been shown to function in the regulation of epithelial cell-cell contacts at adherents junctions as well as in the control of beta-catenin signaling. An increased expression level of this protein was found in the insulin-responsive tissue of obese, insulin-resistant individuals, and may contribute to the pathogenesis of insulin resistance.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
K Tsujikawa et al.
Molecular endocrinology (Baltimore, Md.), 15(2), 271-280 (2001-02-07)
Most receptor-like, transmembrane protein tyrosine phosphatases (PTPases), such as CD45 and the leukocyte common antigen-related (LAR) molecule, have two tandemly repeated PTPase domains in the cytoplasmic segment. The role of each PTPase domain in mediating PTPase activity remains unclear; however
F Ahmad et al.
The Journal of biological chemistry, 272(1), 448-457 (1997-01-03)
The receptor-type protein-tyrosine phosphatase LAR (for leukocyte common antigen-related) has been implicated as a physiological regulator of the insulin receptor. To demonstrate a functional interaction between LAR and the insulin receptor, we incubated CHO cells overexpressing the human insulin receptor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service