Skip to Content
Merck
All Photos(1)

Key Documents

SML3450

Sigma-Aldrich

PA-8

≥95% (HPLC)

Synonym(s):

2-​Amino-​5,​8-​dihydro-​5-​[3-​methoxy-​4-​(2-​propen-​1-​yloxy)​phenyl]​-pyrido[2,​3-​d]​pyrimidine-​4,​7(3H,​6H)​-​dione, PA 8

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N4O4
CAS Number:
Molecular Weight:
342.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

PA-8 is a potent and selective PACAP type I (PAC1) receptor agonist that inhibits the phosphorylation of CREB induced by PACAP in PAC1- receptor-expressing CHO cells. PA-8 blocks PACAP-induced aversive responses and mechanical allodynia in mice.
Potent and selective PACAP type I (PAC1) receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service