Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300069

Sigma-Aldrich

Monoclonal Anti-KLF4 antibody produced in mouse

clone 1E6, ascites fluid

Synonym(s):

EZF, GKLF, KLF4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1E6, monoclonal

mol wt

54 kDa

species reactivity

human

technique(s)

direct ELISA: 1:10,000
immunohistochemistry: 1:200-1:1,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF4(9314)

Immunogen

Purified recombinant fragment of human KLF4 expressed in E.coli.
Mouse monoclonal antibody raised against KLF4

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Claude Fadous-Khalifé et al.
Molecular and clinical oncology, 5(1), 35-40 (2016-06-23)
Lung cancer is most prevalent human cancer worldwide. However, no molecular markers are currently available for predicting lung cancer prognosis. Therefore, identifying novel biomarkers may be useful for improving clinical diagnosis and patient stratification. Krüppel-like factor 4 (KLF4) is a
Jihad Karam et al.
Molecular and clinical oncology, 7(5), 808-814 (2017-11-29)
Late detection and lack of standard treatment strategies in larynx cancer patients result in high levels of mortality and poor prognosis. Prognostic stratification of larynx cancer patients based on molecular prognostic tumor biomarkers may lead to more efficient clinical management.
Qiaojiajie Zhao et al.
The Journal of biological chemistry, 292(6), 2143-2158 (2016-12-29)
The expression of Ring1- and YY1-binding protein (RYBP) is reduced in several human cancers, but the molecular mechanism(s) have remained elusive. In this study, we used human hepatocellular carcinoma (HCC) cell lines and tissue specimens to study the mechanism and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service