Skip to Content
Merck
All Photos(1)

Key Documents

SAB4301208

Sigma-Aldrich

Anti-KDM4C antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GASC1, JHDM3C, JMJD2C, TDRD14C, bA146B14.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

120 kDa

species reactivity

human, mouse

concentration

3.1 mg/mL

technique(s)

western blot: 1:500-1:2000 (Cell Lysate)

isotype

IgG

accession no.

NP_001140166.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KDM4C(23081)

Specificity

The antibody detects endogenous levels of total KDM4C protein.

Immunogen

Synthetic peptide corresponding to a region derived from internal residues of human lysine (K)-specific demethylase 4C

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Kong et al.
Biochemical and biophysical research communications, 514(1), 16-23 (2019-04-25)
One of the hallmark events during liver fibrosis is the transition of quiescent hepatic stellate cells (HSC) into activated myofibroblasts, which are responsible for the production and deposition of pro-fibrogenic proteins. The epigenetic mechanism underlying HSC trans-differentiation is not fully
Meng-Chen Wu et al.
Cell death & disease, 10(2), 68-68 (2019-01-27)
KDM4/JMJD2 Jumonji C-containing histone lysine demethylases (KDM4A-D) constitute an important class of epigenetic modulators in the transcriptional activation of cellular processes and genome stability. Interleukin-8 (IL-8) is overexpressed in gastric cancer, but the mechanisms and particularly the role of the
Nan Li et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(1), 327-338 (2018-07-13)
Liver regeneration is a complicated pathophysiologic process that is regulated by a myriad of signaling pathways and transcription factors. The interaction among these pathways and factors, either cooperatively or antagonistically, may ultimately lead to recovery and restoration of liver function

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service