Skip to Content
Merck
All Photos(2)

Documents

SAB2701826

Sigma-Aldrich

Anti-gapdh antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

cb609, gapd, gapdh, mg:bb02e05

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

zebrafish

technique(s)

immunohistochemistry: suitable
western blot: 5000-10000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

zebrafish ... gapdh(317743)

Immunogen

Recombinant protein containing a sequence corresponding to a region within amino acids 7 and 321 of gapdh according to NP_001108586

Application

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, WB: 1:5000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ran Tao et al.
Oncology letters, 15(4), 5879-5886 (2018-03-20)
There are a limited number of studies reporting on the expression of G protein-coupled receptor kinase 6 (GRK6) in colorectal carcinoma (CRC). The aim of the present study was to investigate and examine the clinical value of GRK6 expression in
Ruijuan Zheng et al.
Nature communications, 9(1), 4072-4072 (2018-10-06)
Tuberculosis (TB) is an infectious disease caused by Mycobacterium tuberculosis (Mtb), and remains a leading public health problem. Previous studies have identified host genetic factors that contribute to Mtb infection outcomes. However, much of the heritability in TB remains unaccounted
Zhe Yi et al.
Oncology letters, 13(3), 1095-1100 (2017-04-30)
The effects of heat shock protein 70 (HSP70)/HSP70-peptide complexes (HSP70-PCs) on the invasion and metastasis ability of hepatocellular carcinoma (HCC) Huh-7 cells were investigated. Wound healing assay revealed that cells treated with HSP70/HSP70-PCs healed faster than negative control cells. HSP70/HSP70-PCs-treated
Ya Zhou et al.
Oncology letters, 16(2), 2645-2653 (2018-07-18)
The incidence of breast cancer is increasing throughout the world. Although significant progress has been made in diagnostic techniques and targeted therapies, the prognosis of breast cancer remains poor. Regulator of differentiation 1 (ROD1) may inhibit the development of several
Feng Liu et al.
Nature communications, 9(1), 4295-4295 (2018-10-18)
Tuberculosis (TB) caused by Mycobacterium tuberculosis (Mtb) kills millions every year, and there is urgent need to develop novel anti-TB agents due to the fast-growing of drug-resistant TB. Although autophagy regulates the intracellular survival of Mtb, the role of calcium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service