Skip to Content
Merck
All Photos(4)

Key Documents

SAB2103783

Sigma-Aldrich

Anti-YBX1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BP-8, Anti-CSDA2, Anti-CSDB, Anti-DBPB, Anti-MDR-NF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

rat, human, rabbit, sheep, horse, bovine, guinea pig, dog, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YBX1(4904)

Immunogen

Synthetic peptide directed towards the middle region of human YBX1

Biochem/physiol Actions

YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. YBX1 binds and stabilizes cytoplasmic mRNA. YBX1 contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. YBX1 binds to promoters that contain a Y-box (5′-CTGATTGGCCAA-3′), such as HLA class II genes. YBX1 regulates the transcription of numerous genes. YBX1 promotes separation of DNA strands that contain mismatches or are modified by cisplatin. YBX1 has endonucleolytic activity and can introduce nicks or breaks into double-stranded DNA (in vitro). YBX1 may play a role in DNA repair. YBX1 is the component of the CRD-mediated complex that promotes MYC mRNA stability.

Sequence

Synthetic peptide located within the following region: NHYRRYPRRRGPPRNYQQNYQNSESGEKNEGSESAPEGQAQQRRPYRRRR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service