Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400688

Sigma-Aldrich

Anti-UPF3A antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-HUPF3A, Anti-RENT3A, Anti-UPF3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UPF3A(65110)

General description

This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The encoded protein is one of two functional homologs to yeast Upf3p. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene is located on the long arm of chromosome 13. Two splice variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

UPF3A (NP_542418.1, 1 a.a. ~ 443 a.a) full-length human protein.

Sequence
MRSEKEGAGGLRAAVAARGPSGREKLSALEVQFHRDSQQQEAETPPTSSSGCGGGAGKPREEKRTALSKVVIRRLPPGLTKEQLEEQLRPLPAHDYFEFFAADLSLYPHLYSRAYINFRNPDDILLFRDRFDGYIFLDSKDPEYKKFLETYCVEEEKTSANPETLLGEMEAKTRELIARRTTPLLEYIKNRKLEKQRIREEKREERRRRELEKKRLREEEKRRRREEERCKKKETDKQKKIAEKEVRIKLLKKPEKGEEPTTEKPKERGEEIDTGGGKQESCAPGAVVKARPMEGSLEEPQETSHSGSDKEHRDVERSQEQESEAQRYHVDDGRRHRAHHEPERLSRRSEDEQRWGKGPGQDRGKKGSQDSGAPGEAMERLGRAQRCDDSPAPRKERLANKDRPALQLYDPGARFRARECGGNRRICKAEGSGTGPEKREEAE

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service