Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400286

Sigma-Aldrich

Anti-HIRA antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DGCR1, Anti-TUP1, Anti-TUPLE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HIRA(7290)

General description

This gene encodes a histone chaperone that preferentially places the variant histone H3.3 in nucleosomes. Orthologs of this gene in yeast, flies, and plants are necessary for the formation of transcriptionally silent heterochomatin. This gene plays an important role in the formation of the senescence-associated heterochromatin foci. These foci likely mediate the irreversible cell cycle changes that occur in senescent cells. It is considered the primary candidate gene in some haploinsufficiency syndromes such as DiGeorge syndrome, and insufficient production of the gene may disrupt normal embryonic development. (provided by RefSeq)

Immunogen

HIRA (NP_003316.3, 1 a.a. ~ 1017 a.a) full-length human protein.

Sequence
MKLLKPTWVNHNGKPIFSVDIHPDGTKFATGGQGQDSGKVVIWNMSPVLQEDDEKDENIPKMLCQMDNHLACVNCVRWSNSGMYLASGGDDKLIMVWKRATYIGPSTVFGSSGKLANVEQWRCVSILRNHSGDVMDVAWSPHDAWLASCSVDNTVVIWNAVKFPEILATLRGHSGLVKGLTWDPVGKYIASQADDRSLKVWRTLDWQLETSITKPFDECGGTTHVLRLSWSPDGHYLVSAHAMNNSGPTAQIIEREGWKTNMDFVGHRKAVTVVKFNPKIFKKKQKNGSSAKPSCPYCCCAVGSKDRSLSVWLTCLKRPLVVIHELFDKSIMDISWTLNGLGILVCSMDGSVAFLDFSQDELGDPLSEEEKSRIHQSTYGKSLAIMTEAQLSTAVIENPEMLKYQRRQQQQQLDQKSAATREMGSATSVAGVVNGESLEDIRKNLLKKQVETRTADGRRRITPLCIAQLDTGDFSTAFFNSIPLSGSLAGTMLSSHSSPQLLPLDSSTPNSFGASKPCTEPVVAASARPAGDSVNKDSMNATSTPAALSPSVLTTPSKIEPMKAFDSRFTERSKATPGAPALTSMTPTAVERLKEQNLVKELRPRDLLESSSDSDEKVPLAKASSLSKRKLELEVETVEKKKKGRPRKDSRLMPVSLSVQSPAALTAEKEAMCLSAPALALKLPIPSPQRAFTLQVSSDPSMYIEVENEVTVVGGVKLSRLKCNREGKEWETVLTSRILTAAGSCDVVCVACEKRMLSVFSTCGRRLLSPILLPSPISTLHCTGSYVMALTAAATLSVWDVHRQVVVVKEESLHSILAGSDMTVSQILLTQHGIPVMNLSDGKAYCFNPSLSTWNLVSDKQDSLAQCADFRSSLPSQDAMLCSGPLAIIQGRTSNSGRQAARLFSVPHVVQQETTLAYLENQVAAALTLQSSHEYRHWLLVYARYLVNEGFEYRLREICKDLLGPVHYSTGSQWESTVVGLRKRELLKELLPVIGQNLRFQRLFTECQEQLDILRDK

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service