Skip to Content
Merck
All Photos(3)

Key Documents

SAB1100050

Sigma-Aldrich

Anti-LDHA (216-228) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-LDH-M, Anti-LDH1, Anti-Lactate dehydrogenase A, Anti-PIG19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LDHA(3939)

General description

Lactate dehydrogenase A (LDHA) or the M subunit is a cytosolic enzyme and is abundantly present in the skeletal muscle. Lactate dehydrogenase B (LDHB) or the H subunit is mainly seen in the heart. Both genes are capable of forming homotetramers or heterotetramers. LDHA gene is located on human chromosome 11p15.4.

Immunogen

synthetic peptide corresponding to amino acids 216-228 of human LDHA

Application

Anti-LDHA (216-228) antibody produced in rabbit has been used in:
  • in situ

  • proximity ligation assay (PLA)
  • immuno-staining
  • western blotting

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

Lactate dehydrogenase A (LDHA) participates in anaerobic and aerobic glycolysis. It can regenerate nicotinamide adenine dinucleotide (NAD+) from NADH to maintain the ATP synthesis in the absence of adequate oxygen. The N-terminus of LDHA is essential for the stability of the structure.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cara J Valvona et al.
Brain pathology (Zurich, Switzerland), 26(1), 3-17 (2015-08-14)
There are over 120 types of brain tumor and approximately 45% of primary brain tumors are gliomas, of which glioblastoma multiforme (GBM) is the most common and aggressive with a median survival rate of 14 months. Despite progress in our
Gloria Bonuccelli et al.
Cell cycle (Georgetown, Tex.), 9(10), 1960-1971 (2010-05-25)
We and others have previously identified a loss of stromal caveolin-1 (Cav-1) in cancer-associated fibroblasts (CAFs) as a powerful single independent predictor of breast cancer patient tumor recurrence, metastasis, tamoxifen-resistance, and poor clinical outcome. However, it remains unknown how loss
Celimene Galiger et al.
BMC cancer, 23(1), 1148-1148 (2023-11-26)
Neuroblastoma (NB), the most common extracranial solid malignancy in children, carries a poor prognosis in high-risk disease, thus requiring novel therapeutic approaches. Survivin is overexpressed in NB, has pro-mitotic and anti-apoptotic functions, and impacts on oxidative phosphorylation (OXPHOS) and aerobic
Veronica Vella et al.
Cells, 8(9) (2019-09-05)
Previously published work has demonstrated that overexpression of the insulin receptor isoform A (IR-A) might play a role in cancer progression and metastasis. The IR has a predominant metabolic role in physiology, but the potential role of IR-A in cancer
Lidia Santos Silva et al.
EMBO reports, 18(12), 2172-2185 (2017-10-27)
Elevated amino acid catabolism is common to many cancers. Here, we show that glioblastoma are excreting large amounts of branched-chain ketoacids (BCKAs), metabolites of branched-chain amino acid (BCAA) catabolism. We show that efflux of BCKAs, as well as pyruvate, is

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service