Skip to Content
Merck
All Photos(1)

Key Documents

S1189

Sigma-Aldrich

Monoclonal Anti-SERCa1 ATPase C-terminus antibody produced in mouse

clone VE121G9, ascites fluid

Synonym(s):

Anti-Sarcoplasmic or Endoplasmic Reticulum Ca2+ ATPase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
WB
clone:
VE121G9, monoclonal
species reactivity:
rabbit, canine, human
citations:
2
technique(s):
indirect immunofluorescence: suitable
western blot: suitable

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

VE121G9, monoclonal

contains

0.05% sodium azide

species reactivity

rabbit, canine, human

technique(s)

indirect immunofluorescence: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP2A1(487)

Immunogen

purified rabbit skeletal muscle sarcoplasmic reticulum.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kálmán Benke et al.
The Journal of heart and lung transplantation : the official publication of the International Society for Heart Transplantation, 40(3), 183-192 (2020-12-06)
Maintenance of cell viability during cold storage is a key issue in organ transplantation. Methane (CH4) bioactivity has recently been recognized in ischemia/reperfusion conditions; we therefore hypothesized that cold storage in CH4-enriched preservation solution can provide an increased defense against
Clara Prats et al.
PloS one, 8(10), e77774-e77774 (2013-11-10)
Skeletal muscle energy metabolism has been a research focus of physiologists for more than a century. Yet, how the use of intramuscular carbohydrate and lipid energy stores are coordinated during different types of exercise remains a subject of debate. Controversy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service