Skip to Content
Merck
All Photos(1)

Documents

RAB0782

Sigma-Aldrich

Mouse Eng / Endoglin ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 16 pg/mL
standard curve range: 16.38-4000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Eng(13805)

General description

The antibody pair provided in this kit recognizes Mouse Endoglin.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matej Vicen et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(5), 6099-6114 (2019-02-13)
Our objective was to investigate the effect of cholesterol [hypercholesterolemia and 7-ketocholesterol (7K)] on endoglin (Eng) expression and regulation with respect to endothelial or vascular dysfunction in vivo and in vitro. In vivo experiments were performed in 2-mo-old atherosclerosis-prone apolipoprotein
F C Brownfoot et al.
Placenta, 35(9), 724-731 (2014-07-22)
Preeclampsia is a serious pregnancy complication. Soluble endoglin (sEng) is released from the placenta and contributes to the maternal endothelial dysfunction seen in preeclampsia. Recently oxysterols, which activate the Liver X Receptor (LXR), have been implicated in producing sEng, by
Joshua M Barnett et al.
Investigative ophthalmology & visual science, 55(10), 6490-6498 (2014-08-28)
We investigated endoglin expression in hypoxic microvascular endothelial cells and retinal endoglin expression in rats that develop experimental oxygen-induced retinopathy (OIR). We also tested neutralizing antibodies (Abs) against endoglin (anti-CD105 Ab) and VEGF (anti-VEGF Ab) either alone or in combination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service