P8147
Platelet-Derived Growth Factor from human platelets
PDGF, lyophilized powder, suitable for cell culture
Synonym(s):
PDGF
About This Item
Recommended Products
biological source
human
Quality Level
Assay
≥95% (SDS-PAGE)
form
lyophilized powder
potency
0.5-10.0 ng/mL EC50
quality
endotoxin tested
mol wt
glycoprotein 28-31 kDa
storage condition
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
impurities
≤1 EU/μg
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... PDGFA(5154) , PDGFB(5155)
General description
Application
Biochem/physiol Actions
Other Notes
Physical form
Analysis Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Platelet-Derived Growth Factor is the principal mitogen found in mammalian serum and is released from platelets during clot formation.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service