Skip to Content
Merck
All Photos(1)

Key Documents

L9168

Sigma-Aldrich

Monoclonal Anti-LKB1 antibody produced in mouse

~1 mg/mL, clone Ley37D/G6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PJS, Anti-STK II (Serine/Threonine Protein Kinase II )

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Ley37D/G6, monoclonal

form

buffered aqueous solution

mol wt

~55 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 2-4 μg/mL using total cell extract of A431 cells

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STK11(6794)

General description

Monoclonal Anti-LKB1 (mouse IgG2b isotype) is derived from the hybridoma Ley37D/G6 produced by the fusion of mouse myeloma cells (p3-NS1/Ag4-1) and splenocytes from BALB/c mice immunized with LKB1 recombinant protein. Liver kinase B1 (LKB1) gene (also termed serine/threonine protein Kinase II STK11 and PJS). LKB1 (55 kDa) is a serine-threonine kinase with highest homology to the cytoplasmic Xenopus protein kinase 1 XEEK1, and weak similarity to many other mammalian protein kinases. It is widely expressed during murine embryonic development, is essential for normal fetal development, and is present in apoptotic intestinal cells.

Immunogen

LKB1 recombinant protein

Application

Monoclonal Anti-LKB1 antibody is suitable for use in immunocytochemistry, immunohistochemistry, immunoprecipitation, western blot (2-4 μg/mL using total cell extract of A431 cells) and indirect ELISA.

Biochem/physiol Actions

Liver kinase B1 (LKB1) induces a G1 cell cycle arrest in tumor cell lines that have lost endogenous LKB1 expression. LKB1 interacts with the chromatin remodeling protein brahma-related gene 1 Brg1, and also with the cell cycle regulatory proteins, LKB1-interacting protein 1 (LIP1) and cyclin-dependent kinase inhibitor p21 (WAF1). It has also been proposed to be involved in wingless (Wnt) signaling and to regulate cell polarity. c-AMP-dependent protein kinase (PKA) phosphorylates LKB1 and it is also prenylated in vivo. LKB1 forms complexes with two proteins STRAD and MO25 that localize LKB1 to the cytoplasm. LKB1 mutations are implicated with Peutz Jeghers cancer syndrome (PJS). LKB1 protein functions as a master upstream kinase of a group of protein kinases named AMPK-related kinases. In lung cancer cells, LKB1 deficiency impairs inhibition of mammalian target of rapamycin (mTOR) activity in response to agents elevating intracellular levels of AMP but not deprivation of growth factor.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Growth arrest by the LKB1 tumor suppressor: induction of p21WAF1/CIP1
Tiainen M, et al.
Human Molecular Genetics, 11(13), 1497-1504 (2002)
LKB1 associates with Brg1 and is necessary for Brg1-induced growth arrest
Marignani PA, et al.
Test, 276(35), 32415-32418 (2001)
A novel de novo mutation in the serine-threonine kinase STK11 gene in a Korean patient with Peutz-Jeghers syndrome
Yoo JH, et al.
BMC Medical Genetics, 9(1), 44-44 (2008)
LKB1 kinase: master and commander of metabolism and polarity
Spicer J and Ashworth A
Current Biology, 14(10), R383-R385 (2004)
LKB1, a novel serine/threonine protein kinase and potential tumour suppressor, is phosphorylated by cAMP-dependent protein kinase (PKA) and prenylated in vivo
COLLINS SP, et al.
The Biochemical Journal, 345(3), 673-680 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service