Documents
EP5490037006
Rotor for Eppendorf® Concentrator Plus
F-40-18-19, non aerosol tight, to hold, 18 x 10 mL flat-bottom tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 18 x 10 mL flat-bottom tubes
to hold 18 x 19 x 66 mm
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490037006 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Concentrator Plus
Legal Information
Eppendorf is a registered trademark of Eppendorf AG