Skip to Content
Merck
All Photos(1)

Key Documents

E7277

Sigma-Aldrich

Elastin, soluble from human lung

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61
Pricing and availability is not currently available.

biological source

human lung

Quality Level

form

lyophilized powder

impurities

salt, free

solubility

H2O: >10 mg/mL (more soluble at pH>7)

UniProt accession no.

storage temp.

2-8°C

InChI

1S/C27H48N6O6/c1-9-17(6)23(32-24(36)19(13-15(2)3)30-18(7)34)26(38)29-14-21(35)31-22(16(4)5)27(39)33-12-10-11-20(33)25(37)28-8/h15-17,19-20,22-23H,9-14H2,1-8H3,(H,28,37)(H,29,38)(H,30,34)(H,31,35)(H,32,36)/t17-,19-,20-,22-,23-/m0/s1

InChI key

DPUYCSDGMSDKKV-MKBYFEBXSA-N

Gene Information

human ... ELN(2006)

Application

Elastin is a structural protein which has been used in studies of chronic obstructive pulmonary disease (COPD). It may be used to investigate why there is a lack of repair for these proteins in patients with COPD.

Preparation Note

Prepared from E7152 using the method of Partrige, et al.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The chemistry of connective tissues. 2. Soluble proteins derived from partial hydrolysis of elastin.
S M PARTRIDGE et al.
The Biochemical journal, 61(1), 11-21 (1955-09-01)
Wei Li et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(5), 1028-1035 (2013-03-16)
Patients with elastin deficiency attributable to gene mutation (supravalvular aortic stenosis) or chromosomal microdeletion (Williams syndrome) are characterized by obstructive arteriopathy resulting from excessive smooth muscle cell (SMC) proliferation, mural expansion, and inadequate vessel size. We investigated whether rapamycin, an
Jae-Yol Lim et al.
PloS one, 8(1), e54256-e54256 (2013-02-02)
Vocal fold (VF) scarring remains a therapeutic challenge. Granulocyte-macrophage colony-stimulating factor (GM-CSF) facilitates epithelial wound healing, and recently, growth factor therapy has been applied to promote tissue repair. This study was undertaken to investigate the effect of GM-CSF on VF
Wolfgang G Mouton et al.
Swiss medical weekly, 143, w13755-w13755 (2013-02-28)
The ultrastructure of venous valves and walls in chronic venous disease was investigated. Consecutive patients were categorised into one of three groups (group A: patients with C1 venous disease in accordance with CEAP (Clinical severity, Etiology, Anatomy, Pathophysiology); group B:
Andrew L Wishart et al.
Science advances, 6(43) (2020-10-23)
The extracellular matrix (ECM), a major component of the tumor microenvironment, promotes local invasion to drive metastasis. Here, we describe a method to study whole-tissue ECM effects from disease states associated with metastasis on tumor cell phenotypes and identify the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service