Skip to Content
Merck
All Photos(1)

Key Documents

E0153

Sigma-Aldrich

Ephrin-A2/Fc Chimera from mouse

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

Ephrin-A2 extracellular domain fused to human IgG1 Fc, Cek7-L, ELF-1, LERK-6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

mouse

Quality Level

recombinant

expressed in NSO cells

Assay

>90% (SDS-PAGE)

form

lyophilized powder

potency

0.16-10 ng/mL

mol wt

monomer calculated mol wt 46 kDa
~57 kDa by SDS-PAGE (reducing)

packaging

pkg of 200 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Efna2(13637)

Biochem/physiol Actions

Member of ephrin ligand family that binds Eph receptor tyrosine kinases (EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, and EphA8). Involved in pattern formation and morphogenesis.

Other Notes

Extracellular domain of mouse ephrin-A2 (amino acids 1-184) is fused to the C-terminal 6X histidine-tagged Fc region of human IgG1.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The biological activity is measured by its ability to bind immobilized recombinant rat EphA5/Fc in ELISA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service