Skip to Content
Merck
All Photos(1)

Key Documents

D3938

Sigma-Aldrich

Anti-DR5 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Death Receptor 5, Anti-KILLER, Anti-TRAIL-R2, Anti-TRICK2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
conjugate:
unconjugated
application:
ARR
WB
clone:
polyclonal
species reactivity:
human
citations:
6
technique(s):
microarray: suitable
western blot: 2 μg/mL using human HeLa and K562 cell extracts

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 57 kDa

species reactivity

human

technique(s)

microarray: suitable
western blot: 2 μg/mL using human HeLa and K562 cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

General description

DR5 is a death domain-containing receptor that is activated through a p53-depedent pathway in response to radiation and DNA damage . DR5 is involved in caspase activation and cell apoptosis . Anti-DR5 antibody is specific for DR5 in humans.

Immunogen

synthetic peptide corresponding to amino acids 388-407 of the human DR5 precursor.

Application

Anti-DR5 antibody may be used in western blot (1 μg/mL using human HeLa and K562 cell extracts), immunoblot and microarray.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongfan Deng et al.
PloS one, 7(5), e37700-e37700 (2012-06-05)
Hepatitis C virus (HCV) is the leading cause of liver fibrosis, cirrhosis and hepatocellular carcinoma. It is believed that continuous liver cell apoptosis contributes to HCV pathogenesis. Recent studies have shown that HCV infection can sensitize host cells to TNF-related
G S Wu et al.
Advances in experimental medicine and biology, 465, 143-151 (2000-05-16)
TRAIL and its emerging receptors are the newest members of the TNF receptor super-family. The activation of TRAIL receptors by ligand binding leads to apoptosis through caspase activation through an as yet unclear signaling pathway that does not require the
R Takimoto et al.
Oncogene, 19(14), 1735-1743 (2000-04-25)
KILLER/DR5, a tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) death receptor gene, has been shown to be induced by DNA damaging agents and radiation in a p53-dependent manner. Although TRAIL is a potential therapeutic agent for cancer, the induction mechanism of
Alena Vaculova et al.
Molecular cancer, 9, 87-87 (2010-04-27)
TRAIL is considered as a promising anti-cancer agent, because of its ability to induce apoptosis in cancer but not in most normal cells. However, growing evidence exist that many cancer cells are resistant to its apoptotic effects. SCLC is a
Vladimira Horova et al.
The FEBS journal, 280(14), 3436-3450 (2013-05-18)
Tumour necrosis factor (TNF) related apoptosis inducing ligand (TRAIL), a membrane-bound ligand from the TNF family, has attracted significant attention due to its rather specific and effective ability to induce apoptotic death in various types of cancer cells via binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service