Documents
CLS766122
Pyrex® pennyhead stopper
stopper joint: 22
Synonym(s):
glass stopper, glass stoppers, stoppers, Corning® 7660
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
material
glass
Joint
joint: stopper22
manufacturer/tradename
Corning
General description
Ground zone length indicates its greatest diameter (mm).
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.