Skip to Content
Merck
All Photos(1)

Key Documents

C1985

Sigma-Aldrich

Anti-Clathrin Light Chain antibody, Mouse monoclonal

clone CON.1, purified from hybridoma cell culture

Synonym(s):

Anti-LCB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CON.1, monoclonal

form

buffered aqueous solution

mol wt

antigen 25-36 kDa (doublet)

species reactivity

human, bovine, mouse, yeast, rat

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1-2 μg/mL using whole rat brain homogenate

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Clathrin, light chain A also known as LCA is a soluble protein composed of heavy and light chain. Clathrin is a protein that plays a major role in the formation of coated vesicles. It coats vesicles from brain, adrenal medulla and non-secreting lymphoma cell line. Clathrin-coated vesicles (CCVs) are major carriers for endocytic cargo and maintain important intracellular trafficking events.

Immunogen

synthetic peptide (amino acids 23-44 in bovine) common to the N-terminus of mammalian clathrin light chains, LCa and LCb, conjugated to KLH. This region is not involved in heavy chain binding and is completely conserved in all mammals.

Application

Monoclonal Anti-Clathrin Light Chain antibody produced in mouse is suitable for the following applications:
  • Immunocytochemistry
  • Immunoprecipitation
  • Microarrary
  • Western blotting at a concentration of 1-2μg/mL using whole rat brain homogenate

Biochem/physiol Actions

Clathrin requires cofactors like ADP-ribosylation factor 6 (ARF6) and adaptin for regulation of receptors. Clathrin light chains (CLCs) are not required for clathrin-mediated endocytosis but plays a critical role for clathrin-mediated trafficking between the TGN and the endosomal system whereas, clathrin heavy chain provides the structural backbone of the clathrin coat. CLCs in mammalian cells function in intracellular membrane trafficking by acting as recruitment proteins for huntingtin-interacting protein 1-related (HIP1R), enabling it to regulate actin assembly on clathrin-coated structures.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delia Bucher et al.
Nature communications, 9(1), 1109-1109 (2018-03-20)
Although essential for many cellular processes, the sequence of structural and molecular events during clathrin-mediated endocytosis remains elusive. While it was long believed that clathrin-coated pits grow with a constant curvature, it was recently suggested that clathrin first assembles to
Viviane Poupon et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(1), 168-173 (2008-01-01)
Clathrin-coated vesicles (CCVs) are major carriers for endocytic cargo and mediate important intracellular trafficking events at the trans-Golgi network (TGN) and endosomes. Whereas clathrin heavy chain provides the structural backbone of the clathrin coat, the role of clathrin light chains
B M Pearse
Proceedings of the National Academy of Sciences of the United States of America, 73(4), 1255-1259 (1976-04-01)
Coated vesicles have been purified from brain, adrenal medulla, and a nonsecreting lymphoma cell line. A single major protein species, clathrin, with an apparent molecular weight of 180,000, forms the coat of all these vesicles. Peptide mapping suggests that the
Martin Hennenberg et al.
The Prostate, 73(7), 715-723 (2013-03-06)
Binding of clathrins or caveolin to G protein-coupled receptors may induce post-translational modifications of receptor function. Receptor regulation by clathrin requires cofactors ADP-ribosylation factor 6 (ARF6) and adaptin, while dynamin is required for clathrin- and caveolin-dependent mechanisms. To investigate the
Masataka Yanagawa et al.
Science signaling, 11(548) (2018-09-20)
G protein-coupled receptors (GPCRs) are major drug targets. Developing a method to measure the activities of GPCRs is essential for pharmacology and drug screening. However, it is difficult to measure the effects of a drug by monitoring the receptor on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service