Skip to Content
Merck
All Photos(2)

Key Documents

AV31964

Sigma-Aldrich

Anti-HOXD3 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Homeobox D3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

guinea pig, rabbit, rat, human, dog, bovine, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXD3(3232)

General description

HoxD3 is a homeobox transcription factor that activates collagen synthesis and angiogenesis and can accelerate wound healing in mice. HOXD3 (HOX4A) has also been implicated in cell adhesion.
Rabbit Anti-HOXD3 (AB1) antibody recognizes canine, human, mouse, rat, and bovine HOXD3.

Immunogen

Synthetic peptide directed towards the middle region of human HOXD3

Application

Rabbit Anti-HOXD3 (AB1) antibody can be used for immunohistochemical assays at a concentration of 4-8μg/ml, using paraffin-embedded tissues. The antibody can also be used for western blot assays 1-2μg/ml.

Biochem/physiol Actions

HOXD3 (homeobox D3) belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that remove the entire HOXD gene cluster or 5′ end of this cluster have been associated with severe limb and genital abnormalities. HOXD3 may play a role in the regulation of cell adhesion processes.

Sequence

Synthetic peptide located within the following region: AHKGAELNGSCMRPGTGNSQGGGGGSQPPGLNSEQQPPQPPPPPPTLPPS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Taniguchi et al.
Blood, 85(10), 2786-2794 (1995-05-15)
We previously isolated the human homeobox gene HOX4A (HOXD3) on chromosome 2 from a human genomic library and determined its nucleotide sequence. In the present study, expression of the HOX4A gene was investigated in human hematopoietic cell lines. Reverse transcriptase-mediated
Scott L Hansen et al.
The American journal of pathology, 163(6), 2421-2431 (2003-11-25)
Poorly healing diabetic wounds are characterized by diminished collagen production and impaired angiogenesis. HoxD3, a homeobox transcription factor that promotes angiogenesis and collagen synthesis, is up-regulated during normal wound repair whereas its expression is diminished in poorly healing wounds of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service