Skip to Content
Merck
All Photos(1)

Key Documents

46069

Sigma-Aldrich

Esterase from horse liver

lyophilized, powder, brown, 0.5-1.0 U/mg

Synonym(s):

HLE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

powder

quality

lyophilized

specific activity

0.5-1.0 U/mg

mol wt

Mr ~70000

color

brown

storage temp.

−20°C

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol ethyl butyrate (Cat. No. 19230) per minute at pH 8.0 and 25°C

Other Notes

Sales restrictions may apply
Slightly higher ee′s can often be obtained compared to PLE-catalysis[1][2][3]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E. Fouque et al.
Synthesis, 661-661 (1989)
L. Blanco et al.
Tetrahedron Letters, 29, 1915-1915 (1988)
J.E. Guibe et al.
Tetrahedron Letters, 30, 67-67 (1989)
B Sànchez-Nogué et al.
Environmental science and pollution research international, 20(5), 3480-3488 (2012-12-06)
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison
Marie C Fortin et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(2), 326-331 (2012-12-12)
Studies on therapeutic drug disposition in humans have shown significant alterations as the result of pregnancy. However, it is not known whether pesticide metabolic capacity changes throughout pregnancy, which could affect exposure of the developing brain. We sought to determine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service