44066
D-Sphingosine
≥98.0% (TLC)
Synonym(s):
Sphingosine, (2S,3R,4E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol
About This Item
Recommended Products
Assay
≥98.0% (TLC)
form
powder
lipid type
sphingolipids
storage temp.
−20°C
SMILES string
OC[C@@](N)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCCC
InChI
1S/C18H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h14-15,17-18,20-21H,2-13,16,19H2,1H3/b15-14+/t17-,18+/m0/s1
InChI key
WWUZIQQURGPMPG-KRWOKUGFSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Packaging
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service