Sodium tungstate is an anti-obesity drug targeting peripheral tissues. In vivo, sodium tungstate reduces body weight gain and food intake through increasing energy expenditure and lipid oxidation, but it also modulates hypothalamic gene expression when orally administered, raising the possibility
Journal of immunotoxicology, 7(3), 174-182 (2010-02-25)
The potential for adverse health effects of using tungsten and its alloys in military munitions are an important concern to both civilians and the US military. The toxicological implications of exposure to tungsten, its alloys, and the soluble tungstate (Na(2)WO(4))
Chemical research in toxicology, 24(4), 488-493 (2011-03-08)
Heavy metal tungsten alloys have replaced lead and depleted uranium in many munitions applications, due to public perception of these elements as environmentally unsafe. Tungsten materials left in the environment may become bioaccessible as tungstate, which might lead to population
It is known that oral administration of sodium tungstate preserves the pancreatic beta cell function in diabetic rats. Healthy and streptozotocin-induced diabetic rats were treated with sodium tungstate for one, three or six weeks, after which the species of W
Tungstate treatment ameliorates experimental diabetes by increasing liver glycogen deposition through an as yet unidentified mechanism. The signalling mechanism of tungstate was studied in CHOIR cells and primary cultured hepatocytes. This compound exerted its pro-glycogenic effects through a new G-protein-dependent