Skip to Content
Merck
All Photos(1)

Key Documents

60458

Sigma-Aldrich

Potassium permanganate

puriss. p.a., ACS reagent, Hg ≤0.000005%, ≥99.0% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
form:
crystals

grade

ACS reagent
puriss. p.a.

Assay

≥99.0% (RT)

form

crystals

impurities

≤0.003% total nitrogen (N)

pH

7.2-9.7 (20 °C, 20 g/L)

anion traces

chloride, chlorate (as Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤10 mg/kg
Ca: ≤150 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤10 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
Hg: ≤0.05 mg/kg
Mg: ≤100 mg/kg
Na: ≤200 mg/kg
Ni: ≤10 mg/kg
Pb: ≤20 mg/kg
Zn: ≤10 mg/kg

SMILES string

[K+].[O-][Mn](=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium permanganate (KMnO4) exhibits strong oxidizing properties. KMnO4 on dissolution in benzene and on complexation with dicyclohexyl-18-crown-6, affords a useful oxidizing reagent. This reagent participates as an oxidant for the oxidation of olefins, alcohols, aldehydes and alkyl benzenes under mild conditions.

Application

Potassium permanganate may be employed as an oxidizing agent in the synthesis of the following:
  • sulfonal (2,2-bisethylsulfonyyl)propane
  • cinchomeronic acid (pyridine-3,4-dicarboxylic acid)
  • isonicotinic acid (pyridine-4-carboxylic acid)
  • manganese(IV) sulfate
Suitable for heterogeneous oxidations.
Used for the decomposition of organic samples for the det. of mercury

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crown polyether chemistry. Potassium permanganate oxidations in benzene.
Sam DJ and Simmons HE.
Journal of the American Chemical Society, 94(11), 4024-4025 (1972)
Eagleson M.
Concise Encyclopedia Chemistry, 1047-1047 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 227-227 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 1097-1097 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 622-622 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service