Skip to Content
Merck
All Photos(1)

Documents

59600

Sigma-Aldrich

3-Methyl-2-butanone

puriss., ≥98.5% (GC)

Synonym(s):

Isopropyl methyl ketone, Methyl isopropyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOCH3
CAS Number:
Molecular Weight:
86.13
Beilstein:
1071238
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Assay

≥98.5% (GC)

form

liquid

expl. lim.

0.34-6.3 %

refractive index

n20/D 1.388 (lit.)
n20/D 1.388

bp

94-95 °C (lit.)

mp

−92 °C (lit.)

solubility

water: soluble(lit.)

density

0.805 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(C)=O

InChI

1S/C5H10O/c1-4(2)5(3)6/h4H,1-3H3

InChI key

SYBYTAAJFKOIEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methyl-2-butanone (Isopropyl methyl ketone, Methyl isopropyl ketone) may be used for the asymmetric synthesis of (R)-3-methyl-2-butanol by Wool-Pd complex catalyzed hydrogenation reaction.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F

Flash Point(C)

-1 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vapor-liquid equilibria of binary and ternary mixtures of cyclohexane, 3-methyl-2-butanone, and octane at 101.3 kPa.
Chen CC, et al.
Journal of Chemical and Engineering Data, 41(3), 557-561 (1996)
Isothermal vapour-liquid equilibria in the binary and ternary systems composed of 2-propanol, 3-methyl-2-butanone and 2, 2, 4-trimethylpentane.
Psutka S and Wichterle I.
Fluid Phase Equilibria, 264(1), 55-61 (2008)
Catalytic behavior of a Wool-Pd complex in asymmetric hydrogenation of diacetone alcohol and 3-methyl-2-butanone.
Yin MO, et al.
J. Mol. Catal. A: Chem., 147(1), 89-92 (1999)
Sebastian Heger et al.
The Science of the total environment, 764, 142902-142902 (2021-03-25)
To avoid potential risks of biofuels on the environment and human, ecotoxicity investigation should be integrated into the early design stage for promising biofuel candidates. In the present study, a green toxicology testing strategy combining experimental bioassays with in silico
Wiley-VCH.
Ullmann's Fine Chemicals (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service