Skip to Content
Merck
All Photos(3)

Key Documents

30323

Sigma-Aldrich

Sodium (meta)periodate

new

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8%

Synonym(s):

Sodium periodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaIO4
CAS Number:
Molecular Weight:
213.89
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24
form:
powder or crystals

grade

ACS reagent
puriss. p.a.

Agency

reag. Ph. Eur.

Assay

≥99.8%

form

powder or crystals

impurities

≤0.001% heavy metals (as Pb)
≤0.01% other halogens (as Cl)

loss

≤0.2% loss on drying, 110 °C

pH

3.5-5.5 (25 °C, 107 g/L)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Fe: ≤10 mg/kg
Mn: ≤1 mg/kg

SMILES string

[Na+].[O-]I(=O)(=O)=O

InChI

1S/HIO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

JQWHASGSAFIOCM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Useful reagent for the oxidation of carbohydrates prior to labeling with biotin hydrazide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1C - STOT RE 1

Target Organs

thymus gland

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation reactions using sodium metaperiodate supported on silica gel.
Gupta DN, et al.
Journal of the Chemical Society. Perkin Transactions 1, 2970-2973 (1981)
Andrew McDowell et al.
Journal of clinical microbiology, 43(1), 326-334 (2005-01-07)
Although two phenotypes of the opportunistic pathogen Propionibacterium acnes (types I and II) have been described, epidemiological investigations of their roles in different infections have not been widely reported. Using immunofluorescence microscopy with monoclonal antibodies (MAbs) QUBPa1 and QUBPa2, specific
Marcelo B Lima et al.
Talanta, 89, 21-26 (2012-01-31)
An automatic method, based on flow-batch (FB), for determining glycerol in biodiesel was developed. The FB systems draw upon the useful features of flow, batch and multi-commutation approaches. The standards and samples preparation, as well as, derivatization and analysis were
Dmitriy Berillo et al.
Macromolecular bioscience, 12(8), 1090-1099 (2012-06-08)
Macroporous scaffolds composed of chitosan and using oxidized dextran as a crosslinker are produced through cryogelation. Introducing gelatin as a third component into the structure results in the formation of mesopores in the pore walls, which are not seen if
Abeer Al-Ghouleh et al.
PloS one, 7(3), e33929-e33929 (2012-04-06)
Allergens are initiators of both innate and adaptive immune responses. They are recognised at the site of entry by epithelial and dendritic cells (DCs), both of which activate innate inflammatory circuits that can collectively induce Th2 immune responses. In an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service