Skip to Content
Merck
All Photos(1)

Key Documents

Y0000322

Loperamide hydrochloride for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Loperamide hydrochloride, 4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutyramide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H33ClN2O2 · HCl
CAS Number:
Molecular Weight:
513.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

loperamide

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN(C)C(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4

InChI

1S/C29H33ClN2O2.ClH/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32-20-17-28(34,18-21-32)23-13-15-26(30)16-14-23;/h3-16,34H,17-22H2,1-2H3;1H

InChI key

PGYPOBZJRVSMDS-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Loperamide hydrochloride for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Ringkamp et al.
PloS one, 7(7), e42105-e42105 (2012-08-01)
Loperamide reverses signs of mechanical hypersensitivity in an animal model of neuropathic pain suggesting that peripheral opioid receptors may be suitable targets for the treatment of neuropathic pain. Since little is known about loperamide effects on the responsiveness of primary
Arnold Wald
Best practice & research. Clinical gastroenterology, 26(5), 573-580 (2013-02-07)
IBS is a functional gastrointestinal disorder which has been subtyped according to bowel habits. This review presents recommendations for IBS-D which makes up about 1/3 of all patients and which is defined as IBS with loose or watery stools with
Paul A Boakye et al.
Journal of ethnopharmacology, 142(2), 539-547 (2012-05-31)
The extract from the stem bark of Garcinia buchananii trees is used as an anti-diarrhea remedy in sub-Saharan Africa. We tested the hypothesis that G. buchananii bark extract and its anti-motility fractions are effective treatments against lactose-induced diarrhea. A high-lactose
Flavia Neri et al.
Journal of biomedicine & biotechnology, 2012, 896162-896162 (2012-06-01)
Medical treatment in chronic constipation is not always successful. Surgery is indicated in unresponsive selected severe cases. This study presents the distal venous colic ligation in rat as a novel surgical approach. 16 rats (study group) were evaluated in 3
Kai-Chun Cheng et al.
Pharmacology & therapeutics, 139(3), 334-340 (2013-05-22)
Type-2 diabetes is one of the fastest growing public health problems worldwide resulting from both environmental and genetic factors. Activation of μ-opioid receptor (MOR) could result in reversal of the impairment of insulin-stimulated glucose disposal in genetically obese Zucker rats

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service