Skip to Content
Merck
All Photos(1)

Key Documents

PHR1194

Supelco

Verapamil Related Compound E

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

3,4-Dimethoxybenzaldehyde, Methylvanillin, NSC 24521, NSC 8500, Vanillin methyl ether, Veratraldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CHO
CAS Number:
Molecular Weight:
166.17
Beilstein:
473899
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1711439

API family

verapamil

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

281 °C (lit.)

mp

40-43 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[H]C(=O)c1ccc(OC)c(OC)c1

InChI

1S/C9H10O3/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-6H,1-2H3

InChI key

WJUFSDZVCOTFON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9010 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Suresh et al.
Plant physiology and biochemistry : PPB, 43(2), 125-131 (2005-04-12)
Normal root cultures of Capsicum frutescens biotransform externally fed precursors, like caffeic acid and veratraldehyde, to vanillin and other related metabolites. The bioconversion of caffeic acid to further metabolites--viz. vanillin, vanillylamine, vanillic acid--was shown to be elicited by treating the
Francesca D'Acunzo et al.
Biochemical and biophysical research communications, 313(1), 17-21 (2003-12-16)
Veratrylchitosan, a polysaccharide-supported lignin model compound, has been synthesised by covalently attaching 3-(3,4-dimethoxybenzyloxy)propionic acid to the polysaccharide chitosan through an amide linkage. When this polymer was used as a substrate in the oxidation promoted by lignin peroxidase (LiP), significant decomposition
J Kavitha et al.
Journal of Asian natural products research, 2(2), 157-160 (2001-03-17)
N-[3-(3,4-Dimethoxyphenyl)propanoyl]pyrrole (1) has been synthesized in three steps starting from veratraldehyde (2) with an overall yield of 66%.
V Elisashvili et al.
Prikladnaia biokhimiia i mikrobiologiia, 38(3), 243-247 (2002-06-19)
The activities of carboxymethylcellulase and xylanase in the higher basidial fungus Cerrena unicolor grown in avicel-containing medium reached 1.95 and 1.50 units per mg protein, respectively, whereas in mannitol-containing medium they ranged from 0.02 to 0.05 units per mg protein.
Troy M Larson et al.
Biotechnology letters, 35(2), 225-231 (2012-11-08)
Laccases play an important role in the biological break down of lignin and have great potential in the deconstruction of lignocellulosic feedstocks. We examined 16 laccases, both commercially prepared and crude extracts, for their ability to oxidize veratryl alcohol in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service