Skip to Content
Merck
All Photos(1)

Key Documents

P3640000

Propyl gallate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3,4,5-Trihydroxybenzoic acid propyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
Beilstein:
1877976
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

propyl gallate

manufacturer/tradename

EDQM

mp

146-149 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3

InChI key

ZTHYODDOHIVTJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Reconstitution

Prepare a 0.1M solution in glycerol:PBS (9:1)

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

368.6 °F - closed cup

Flash Point(C)

187 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasunobu Kawano et al.
Resuscitation, 83(2), 249-252 (2011-08-02)
The present study was conducted to assess the effects of intraperitoneal administration of n-propyl gallate (PG) on hippocampal neuronal survival after forebrain ischemia. Forty male Sprague-Dawley rats were randomly assigned to one of 6 groups. Animals in the PG-I-10, PG-I-8
Paola Colindres et al.
Journal of the science of food and agriculture, 91(5), 963-968 (2011-01-22)
The effect of selected antioxidants (grape seed extract (GS), oleoresin rosemary (OR), water-soluble oregano extract (WO), propyl gallate (PG), butylated hydroxyanisole (BHA)), butylated hydroxytoluene (BHT)) on sensory, color and oxidative stability of cooked, frozen, reheated ground beef patties was evaluated.
Sunil Yeruva et al.
Frontiers in physiology, 11, 430-430 (2020-06-09)
Intercalated discs (ICDs), which connect adjacent cardiomyocytes, are composed of desmosomes, adherens junctions (AJs) and gap junctions (GJs). Previous data demonstrated that adrenergic signaling enhances cardiac myocyte cohesion, referred to as positive adhesiotropy, via PKA-mediated phosphorylation of plakoglobin (PG). However
Meixia Huang et al.
Journal of pharmaceutical and biomedical analysis, 58, 12-18 (2011-10-08)
The purpose of this study was to prepare a propyl gallate (PrG) molecular imprinted polymer as a cartridge stuffing material to isolate antiplatelet active ingredients. A macroporous polymer was synthesized utilizing ethylene glycol dimethacrylate (EDMA) as the crosslinking agent, PrG
Kanako Inoue et al.
FEMS microbiology letters, 326(1), 83-90 (2011-11-19)
Azoxystrobin (AZ), a strobilurin-derived fungicide, is known to inhibit mitochondrial respiration in fungi by blocking the electron transport chain in the inner mitochondrial membrane. Germination was strongly inhibited when Botrytis cinerea spore suspension was treated with AZ and the alternative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service