Skip to Content
Merck
All Photos(1)

Key Documents

79737

Supelco

Hederacoside C

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H96O26
CAS Number:
Molecular Weight:
1221.38
Beilstein:
4229541
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

mp

222 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

[H][C@]12CC=C3C4CC(C)(C)CC[C@@]4(CC[C@@]3(C)[C@]1(C)CCC5[C@](C)(CO)[C@H](CC[C@]25C)O[C@@H]6OC[C@H](O)[C@H](O)[C@H]6O[C@@H]7O[C@@H](C)[C@H](O)[C@@H](O)[C@H]7O)C(=O)O[C@@H]8O[C@H](CO[C@@H]9O[C@H](CO)[C@@H](O[C@@H]%10O[C@@H](C)[C@H](O)[C@@H](O)[C@H]%10O)[C@H](O)[C@H]9O)[C@@H](O)[C@H](O)[C@H]8O

InChI

1S/C59H96O26/c1-24-34(63)38(67)42(71)49(78-24)83-46-29(20-60)80-48(45(74)41(46)70)77-22-30-37(66)40(69)44(73)51(81-30)85-53(75)59-17-15-54(3,4)19-27(59)26-9-10-32-55(5)13-12-33(56(6,23-61)31(55)11-14-58(32,8)57(26,7)16-18-59)82-52-47(36(65)28(62)21-76-52)84-50-43(72)39(68)35(64)25(2)79-50/h9,24-25,27-52,60-74H,10-23H2,1-8H3/t24-,25-,27?,28-,29+,30+,31?,32+,33-,34-,35-,36-,37+,38+,39+,40-,41+,42+,43+,44+,45+,46+,47+,48+,49-,50-,51-,52-,55-,56-,57+,58+,59-/m0/s1

InChI key

RYHDIBJJJRNDSX-WBOYGZCJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hederacoside C is a major bioactive constituent isolated from Hedera helix L. It exhibits pharmacological properties such as antibacterial, expectorant, bronchodilator and bronchospasmolytic effects. It is widely used in the treatment of respiratory disorders involving acute respiratory infections, chronic inflammatory bronchitis and productive coughs.[1]

Application

Hederacoside C may be used as an analytical reference standard for the quantification of the analyte in Ivy leaf extract[2][1] using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: hedera

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

An ultra-high-performance liquid chromatography-tandem mass spectrometric method for the determination of hederacoside C, a drug candidate for respiratory disorder, in rat plasma
Rehman US, et al.
Journal of Pharmaceutical and Biomedical Analysis, 129, 90-95 (2016)
Determination of saponins and flavonoids in ivy leaf extracts using HPLC-DA
Yu M, et al.
Journal of Chromatographic Science, 53(4), 478-483 (2014)
Eun-Ha Joh et al.
Phytotherapy research : PTR, 26(4), 546-551 (2011-09-20)
The stem-bark of Kalopanax pictus (KP, family Araliaceae), which contains triterpenoid saponins, has been shown to exhibit anticarcinogenic, antiinflammatory, antirheumatoid and antidiabetic activities. In a preliminary study, a KP methanol extract demonstrated acetylcholinesterase activity in vitro and memory enhancement in
D H Kim et al.
Biological & pharmaceutical bulletin, 21(4), 360-365 (1998-05-20)
To investigate the relationship between the intestinal bacterial metabolism of kalopanaxsaponin B and H from Kalopanax pictus (Araliaceae), and their antidiabetic effect, kalopanaxsaponin B and H were metabolized by human intestinal microflora and the antidiabetic activity of their metabolites was
S H Cho et al.
Archives of pharmacal research, 14(1), 19-24 (1991-03-01)
One new triterpenoidal saponin, saponin F(2) has been isolated from the bark of Kalopanax pictum Nakai var. typicum (Araliaceae), together with one known saponin, kizuta saponin K12 (1). On the basis of chemico-spectral evidences, the structure of 2 has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service