Skip to Content
Merck
All Photos(1)

Documents

78278

Supelco

Mepiquat iodide-(methyl-d3)

PESTANAL®, analytical standard

Synonym(s):

1-Methyl-1-methyl-d3-piperidinium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D3H13IN
CAS Number:
Molecular Weight:
244.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

C[N+]1(C)CCCCC1.[I-]

InChI

1S/C7H16N.HI/c1-8(2)6-4-3-5-7-8;/h3-7H2,1-2H3;1H/q+1;/p-1

InChI key

HSQOPVUSHBNOOL-UHFFFAOYSA-M

Related Categories

General description

Mepiquat iodide-(methyl-d3) is an iodide salt of mepiquat which is widely used as a plant growth regulator in agriculture.

Application

Mepiquat iodide-(methyl-d3) may be used as an internal standard for the determination of mepiquat in oven-cooked beef by high performance liquid chromatography (HPLC-MS) using electrospray ionization (ESI) in the positive ion mode and in cooked vegetables and food products by LC-MS/MS analysis.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Growth retardants: effects on gibberellin biosynthesis and other metabolic pathways.
Rademacher, et al.
Annual Review of Plant Biology, 51(1), 501-531 (2000)
Heat-induced formation of mepiquat by decarboxylation of pipecolic acid and its betaine derivative. Part 2: Natural formation in cooked vegetables and selected food products
Yuan Y, et al.
Food Chemistry, 228, 99-105 (2017)
Carnitine, a New Precursor in the Formation of the Plant Growth Regulator Mepiquat.
Yuan, et al.
Journal of Agricultural and Food Chemistry, 51(1), 501-531 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service