Skip to Content
Merck
All Photos(1)

Documents

67226

Supelco

Tin (impurities)

manufactured by MBH Analytical Ltd (71X SR0)

Synonym(s):

Tin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

material

disks

grade

analytical standard

resistivity

11 μΩ-cm, 20°C

technique(s)

XRF: suitable

diam. × H

40 mm × 15 mm , dimensions are approximate

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

Analysis Note

Values of analytes vary lot to lot.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason D Sagers et al.
The Journal of the Acoustical Society of America, 132(4), 2173-2183 (2012-10-09)
Thomas Edison's phonograph was a landmark acoustic invention. In this paper, the phonograph is presented as a tool for education in acoustics. A brief history of the phonograph is outlined and an analogous circuit model that describes its dynamic response
Jeremy Ball et al.
Optics express, 20(18), 20266-20275 (2012-10-06)
The optical characteristics of silicon nanowires grown on Si layers on glass have been modeled using the FDTD (Finite Difference Time Domain) technique and compared with experimental results. The wires were grown by the VLS (vapour-liquid-solid) method using Sn catalyst
Bing Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(41), 16459-16462 (2012-09-27)
Using synchrotron high-pressure X-ray diffraction at cryogenic temperatures, we have established the phase diagram for calcium up to 110 GPa and 5-300 K. We discovered the long-sought for theoretically predicted β-tin structured calcium with I4(1)/amd symmetry at 35 GPa in
Qian Cheng et al.
ChemSusChem, 6(1), 102-109 (2013-01-03)
Spray pyrolysis was used to prepare films of AgInS(2) (AIS) with and without Sn as an extrinsic dopant. The photoelectrochemical performance of these films was evaluated after annealing under a N(2) or S atmosphere with different amounts of the Sn
Olga G Tsay et al.
Inorganic chemistry, 52(4), 1991-1999 (2013-02-05)
Various R/Ar-functionalized tin 5,10,15-tris(pentafluorophenyl)corrolate derivatives are reported herein including the first ferrocenyltin corrolate species. The isopropyl, sec-butyl-, 2-methyl-n-butyl-, phenyl-, 2-thienyl-, and ferrocenyltin species have been prepared and characterized through (1)H, (13)C, and (119)Sn HMQC NMR spectroscopy, mass spectrometry, UV-vis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service