Skip to Content
Merck
All Photos(1)

Documents

61076

Supelco

DL-4-Hydroxy-2-ketoglutaric acid lithium salt

analytical standard

Synonym(s):

4-Hydroxy-2-oxoglutaric acid lithium salt, DL-KHG Li

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O6 · xLi+
CAS Number:
Molecular Weight:
162.10 (free acid basis)
Beilstein:
1780096
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

−20°C

InChI

1S/C5H6O6/c6-2(4(8)9)1-3(7)5(10)11/h2,6H,1H2,(H,8,9)(H,10,11)

InChI key

WXSKVKPSMAHCSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-4-Hydroxy-2-ketoglutaric acid lithium salt (HOG) is a precursor substrate of 4-hydroxy-2-oxoglutarate aldolase (HOGA1), a unique enzyme in the hydroxyproline catabolism pathway. It is reported that the primary hyperoxaluria type 3, which is a recently discovered form of the autosomal recessive disease, results from the mutations in the gene coding for HOGA1.

Application

DL-4-Hydroxy-2-ketoglutaric acid lithium salt may be used as an analytical reference standard for the determination of the analyte in animal biological samples using ion chromatography coupled to negative electrospray mass spectrometry (IC/ES-MS). This can be used to determine the primary hyperoxalurias PH Type 3 (PH3), caused by the mutations in the mitochondrial enzyme, 4-hydroxy-2-oxoglutarate aldolase (HOGA).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroxyproline metabolism in a mouse model of Primary Hyperoxaluria Type 3
Li X, et al.
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease, 1852(12), 2700-2705 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service