Skip to Content
Merck
All Photos(1)

Key Documents

523291

Supelco

Poly(2-vinylpyridine)

analytical standard, average Mw 5,000 (Typical), average Mn 4,800 (Typical)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

mol wt

average Mn 4,800 (Typical)
average Mw 5,000 (Typical)

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

InChI

1S/C7H7N/c1-2-7-5-3-4-6-8-7/h2-6H,1H2

InChI key

KGIGUEBEKRSTEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keewook Paeng et al.
The Journal of chemical physics, 134(2), 024901-024901 (2011-01-19)
A temperature-ramping anisotropy measurement is introduced as an efficient way to study molecular motion in polymer glasses. For these experiments, fluorescent molecules were dispersed in the polymer glass and the reorientation of these dyes was used as a probe of
Tianying Jiang et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 24(37), 375109-375109 (2012-08-24)
Here we investigate the flow properties of suspensions of dicolloidal particles composed of interpenetrating spheres where one sphere is rich in polystyrene and the second is rich in poly 2-vinyl pyridine. The synthesis method is designed to create both anisotropic
Weihua Tao et al.
Journal of hazardous materials, 175(1-3), 111-116 (2009-11-03)
In the present study, the performance, behavior and mechanism of synthetic poly(2-vinylpyridine) resin (WH-225) adsorbing 5-sodiosulfoisophthalic acids (SIPA) from the aqueous solution were investigated, and two commercial adsorbents, namely, hypercrosslinked adsorbent NDA-100 and macroporous adsorbent XAD-4 were employed as reference.
Mohammad Changez et al.
Advanced materials (Deerfield Beach, Fla.), 24(24), 3253-3257 (2012-05-19)
The reaction between atactic poly(2-vinylpyridine) and 1,4-dibromobutane leads to formation of long-range 3D molecular ordering in polymer chains mainly because the side group (pyridine) of the polymer chain changes to a syndotactic configuration. This may enable the production of functional
Andreas Walther et al.
Chemical communications (Cambridge, England), (9)(9), 1127-1129 (2009-02-20)
We report on the unexpected finding of the formation of hydrophobic bridges between polybutadiene patches of multicompartment micelles based on amphiphilic miktoarm star terpolymers in water.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service