Skip to Content
Merck
All Photos(1)

Key Documents

50292

Supelco

Aloin

analytical standard

Synonym(s):

1,8-Dihydroxy-10-(β-D-glucopyranosyl)-3-(hydroxymethyl)-9(10H)-anthracenone, 10-β-D-Glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthracenone, Aloin A, Barbaloin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O9
CAS Number:
Molecular Weight:
418.39
Beilstein:
6077558
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water (Karl Fischer)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@H]2c3cccc(O)c3C(=O)c4c(O)cc(CO)cc24

InChI

1S/C21H22O9/c22-6-8-4-10-14(21-20(29)19(28)17(26)13(7-23)30-21)9-2-1-3-11(24)15(9)18(27)16(10)12(25)5-8/h1-5,13-14,17,19-26,28-29H,6-7H2/t13-,14+,17-,19+,20-,21+/m1/s1

InChI key

AFHJQYHRLPMKHU-OSYMLPPYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Purgative activity in rats requires activation by Eubacterium sp. strain BAR or similar intestinal anaerobe, presumably by conversion to aloe-emodin anthrone.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Ishii et al.
Biological & pharmaceutical bulletin, 17(5), 651-653 (1994-05-01)
Aloe-emodin-9-anthrone(AE-anthrone), produced from barbaloin in the rat large intestine, caused not only an increase in the intestinal water content but also stimulated mucus secretion. This might play an important role in the occurrence of diarrhea. It was demonstrated that the
Hong-Xing Wang et al.
Ying yong sheng tai xue bao = The journal of applied ecology, 21(1), 260-264 (2010-04-15)
By using transmission electron microscopy and high performance liquid chromatography, this paper studied the effects of enhanced UV-B radiation on the leaf anthraquinones content and cell ultrastructure of Aloe vera L. After treated with enhanced UV-B radiation 6 hours per
Y Ishii et al.
Biological & pharmaceutical bulletin, 16(10), 1040-1040 (1993-10-01)
As individual differences in sensitivity in the laxative activity of barbaloin were observed in rats, a selection test for barbaloin (31.1 mg/kg, p.o.) was carried out between 1989 and 1991. The annual percentages of rats exhibiting a reaction positive to
M Hattori et al.
Pharmacology, 47 Suppl 1, 125-133 (1993-10-01)
A strictly anaerobic bacterium, Bifidobacterium sp. SEN, capable of hydrolyzing the O-glucosyl of sennosides was isolated from human feces. The bacterium stepwisely hydrolyzed sennoside B to sennidin B through sennidin-8-monoglucoside in PYF medium but not in GAM broth. Addition of
Jianniao Tian et al.
Chemical & pharmaceutical bulletin, 51(5), 579-582 (2003-05-09)
The fluorescence quenching reactions of barbaloin with bovine serum albumin (BSA) in pH 7.20 Tris-HCl buffer solution were studied. The quenching mechanism of BSA by barbaloin was interpreted using the Stern-Volmer (S-V) mechanism. The binding constant K values were 2.78

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service