Skip to Content
Merck
All Photos(1)

Key Documents

46565

Supelco

Estriol

VETRANAL®, analytical standard

Synonym(s):

1,3,5(10)-Estratriene-3,16α,17β-triol, 16α-Hydroxyestradiol, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
Beilstein:
2508172
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

280-282 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O

InChI

1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1

InChI key

PROQIPRRNZUXQM-ZXXIGWHRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina Sneitz et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(3), 582-591 (2013-01-05)
The glucuronidation of estriol, 16-epiestriol, and 17-epiestriol by the human UDP-glucuronosyltransferases (UGTs) of subfamilies 1A, 2A, and 2B was examined. UGT1A10 is highly active in the conjugation of the 3-OH in all these estriols, whereas UGT2B7 is the most active
J J Evans et al.
Clinical chemistry, 30(1), 120-121 (1984-01-01)
Saliva would have advantages over plasma or urine for monitoring estriol during pregnancy. Specimen collection, after stimulation of flow by citric acid, is non-invasive and simple. We measured concentrations of unconjugated estriol in saliva and compared them with those in
E Diczfalusy
Journal of steroid biochemistry, 20(4B), 945-953 (1984-04-01)
The early history of estriol is reviewed with special emphasis on its isolation, identification, quantitation in tissues and body fluids and its unique biogenesis in the human feto-placental unit. The relationship to epimeric estriols and acidic estrogens and the pharmacokinetic
Marcelo O Dietrich et al.
Trends in neurosciences, 36(2), 65-73 (2013-01-16)
The past 20 years witnessed an enormous leap in understanding of the central regulation of whole-body energy metabolism. Genetic tools have enabled identification of the region-specific expression of peripheral metabolic hormone receptors and have identified neuronal circuits that mediate the
G P Vooijs et al.
European journal of obstetrics, gynecology, and reproductive biology, 62(1), 101-106 (1995-09-01)
To gain more insight into whether intravaginal treatment of local urogenital complaints with the mild-acting oestrogen estriol is capable of inducing proliferation of the endometrium, the results of the clinical studies that have been published over the years have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service