Skip to Content
Merck
All Photos(1)

Key Documents

37013

Supelco

Moniliformin sodium salt solution

~100 μg/mL in acetonitrile: water, analytical standard

Synonym(s):

1-Hydroxycyclobut-1-ene-3,4-dione sodium salt, Semisquaric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4HNaO3
CAS Number:
Molecular Weight:
120.04
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~100 μg/mL in acetonitrile: water

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

O=C1C(C=C1O[Na])=O

InChI

1S/C4H2O3.Na/c5-2-1-3(6)4(2)7;/h1,5H;/q;+1/p-1

InChI key

FERDNJVXTWPNSA-UHFFFAOYSA-M

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥92.5% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Walburga von Bargen et al.
Journal of agricultural and food chemistry, 60(14), 3586-3591 (2012-03-21)
Moniliformin is a mycotoxin produced by fungi of the Fusarium genus and occurs as a contaminant of different cereals worldwide. This study describes the first application of isotopically labeled (13)C(2)-moniliformin for the analysis of moniliformin in cereals. Moniliformin is a
An Zhang et al.
Journal of Zhejiang University. Science. B, 11(3), 200-208 (2010-03-06)
To investigate the effects of mycotoxin moniliformin (MON) on the metabolism of aggrecan and type II collagen in human chondrocytes in vitro and the relationship between MON and Kashin-Beck disease (KBD). Human chondrocytes were isolated and cultured on bone matrix
A Waskiewicz et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 27(5), 608-615 (2010-05-11)
The principal aim of this study was to estimate the formation of fumonisins (FB(1) and FB(2)), moniliformin (MON), and ergosterol (ERG) by Fusarium oxysporum and Fusarium proliferatum, while the formation of beauvericin (BEA) was estimated by the latter Fusarium species
R Kristensen et al.
Journal of applied microbiology, 102(4), 1071-1081 (2007-03-27)
To develop a multiplex identification method for trichothecene- and moniliformin-producing Fusarium species. In this article, we present a single nucleotide polymorphism (SNP) assay to simultaneously detect and identify 16 trichothecene- and moniliformin-producing Fusarium species. A number of SNP primers are
Silvio Uhlig et al.
International journal of food microbiology, 119(1-2), 17-24 (2007-09-22)
The Fusarium species complex found on small-grain cereals in Northern Europe is largely dominated by F. avenaceum, while other important species include F. tricinctum, F. poae, F. culmorum and F. graminearum. The dominance of F. avenaceum has in recent years

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service