Skip to Content
Merck
All Photos(1)

Key Documents

36698

Supelco

Cumene

PESTANAL®

Synonym(s):

(1-Methylethyl)benzene, 2-Phenylpropane, Isopropylbenzene, NSC 8776

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)2
CAS Number:
Molecular Weight:
120.19
Beilstein:
1236613
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

vapor density

4.1 (vs air)

Quality Level

vapor pressure

8 mmHg ( 20 °C)
9.7 mmHg ( 37.7 °C)

product line

PESTANAL®

autoignition temp.

797 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.491 (lit.)

bp

152-154 °C (lit.)

mp

−96 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)c1ccccc1

InChI

1S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8H,1-2H3

InChI key

RWGFKTVRMDUZSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cumene is an organic compound obtained, in the process of acid-catalyzed alkylation of benzene with propylene.[1]

Application

Cumene may be used as a reference standard for the determination of cumene in water samples by gas chromatography-mass spectrometry method (GC-MS). Cumene may be used as an analytical reference standard for the quantification of the analyte in environmental samples using gas chromatography technique.[2]
Cumene may be used as a reference standard for the determination of cumene in water samples by gas chromatography-mass spectrometry method (GC-MS).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of toxic toluene, xylene and cumene in different lake waters.
Amzad H M, et al.
International Journal of Environmental Research, 4(2), 341-346 (2010)
Toxicity assessment of atrazine and related triazine compounds in the microtox assay, and computational modeling for their structure-activity relationship
Tchounwou.BP, et al.
International Journal of Molecular Sciences, 1(4), 63-74 (2000)
Use of solid-phase extraction in the determination of benzene, toluene, ethylbenzene, xylene and cumene in spiked soil and investigation of soil spiking methods
Meney.MK, et al.
Analyst, 123(2), 195-2000 (1998)
Sibylle Schliemann et al.
Contact dermatitis, 63(6), 347-356 (2010-11-03)
Skin exposure to lipophilic irritants at workplaces is a risk factor for the development of occupational irritant hand dermatitis. Suitable lipophilic model irritants to be used in repeated irritation studies in volunteers are needed to gain a better understanding of
Sibylle Schliemann et al.
Contact dermatitis, 65(2), 107-114 (2011-06-23)
Neurosensory irritation is a subjective phenomenon induced by a number of chemicals. Objectives. To investigate the sensory irritation induced by two organic solvents - n-octane and cumene - between two groups of volunteers, stingers and non-stingers, identified as such according

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service