Skip to Content
Merck
All Photos(1)

Key Documents

36536

Supelco

Tralkoxydim

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27NO3
CAS Number:
Molecular Weight:
329.43
Beilstein:
8577523
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCO\N=C(/CC)C1=C(O)CC(CC1=O)c2c(C)cc(C)cc2C

InChI

1S/C20H27NO3/c1-6-16(21-24-7-2)20-17(22)10-15(11-18(20)23)19-13(4)8-12(3)9-14(19)5/h8-9,15,22H,6-7,10-11H2,1-5H3/b21-16+

InChI key

DQFPEYARZIQXRM-LTGZKZEYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tralkoxydim belongs to the class of cyclohexanedione herbicides, used for the control of many monocotyledons species.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tralkoxydim may be used as an analytical reference standard for the quantification of the analyte in environmental samples and food products using various chromatographic techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of acetyl-CoA carboxylase activity by haloxyfop and tralkoxydim
Secor J and Cseke C
Plant Physiology, 86(1), 10-12 (1988)
A M Brady et al.
Toxicology, 84(1-3), 199-215 (1993-11-12)
2-[1-(Ethoxyimino)propyl]-3-hydroxy-5-(2,4,6-trimethylphenyl) cyclohex-2-enone (ETC) is a novel alkyl ketone herbicide. Continuous administration of ETC to mice for 28 days resulted in marked liver enlargement and severe intrahepatic cholestasis. These effects have been shown to result directly from a rapid and marked
Application of SPE-HPLC-DAD and SPE-TLC-DAD to the determination of pesticides in real water samples.
Tuzimski T.
Journal of Separation Science, 31(20), 3537-3542 (2008)
Determination of 142 pesticides in fruit-and vegetable-based infant foods by liquid chromatography/electrospray ionization-tandem mass spectrometry and estimation of measurement uncertainty.
Wang J and Leung D
Journal of AOAC (Association of Official Analytical Chemists) International, 92(1), 279-301 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service