Skip to Content
Merck
All Photos(2)

Documents

36199

Supelco

Tridemorph

PESTANAL®, analytical standard

Synonym(s):

2,6-Dimethyl-4-tridecylmorpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H39NO
CAS Number:
Molecular Weight:
297.52
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCCN1CC(C)OC(C)C1

InChI

1S/C19H39NO/c1-4-5-6-7-8-9-10-11-12-13-14-15-20-16-18(2)21-19(3)17-20/h18-19H,4-17H2,1-3H3

InChI key

YTOPFCCWCSOHFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Zamora et al.
Journal of chromatography. A, 1045(1-2), 137-143 (2004-09-24)
A rapid and sensitive liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS-MS) method for the determination of tridemorph and other pre- and post-harvest fungicides (carbendazim, thiabendazole, imazalil, propiconazole and bitertanol) in banana and orange samples has been developed and validated. The sample
Yandu Lu et al.
mSystems, 5(5) (2020-10-01)
Cnidarians cannot synthesize sterols (which play essential roles in growth and development) de novo but often use sterols acquired from endosymbiotic dinoflagellates. While sterol availability can impact the mutualistic interaction between coral host and algal symbiont, the biosynthetic pathways (in
A Rahier et al.
Biochemical Society transactions, 11(5), 537-543 (1983-10-01)
25-Azacycloartanol (I), 2-aza-2-dihydrosqualene (II) and Tridemorph (2,6-dimethyl-N-tridecylmorpholine) (III) are potent inhibitors of higher plant sterol biosynthesis. The first two molecules have been designed using rational enzymological concepts. I, II and III were shown to inhibit the S-adenosyl-L-methionine: cycloartenol C-24-methyltransferase, the
Xia Tian et al.
The Journal of organic chemistry, 71(8), 3176-3183 (2006-04-08)
Tandem Prins cyclization and Friedel-Crafts reaction with an electron-rich aromatic ring were used to prepare the core structures of calyxin natural products. The proposed structure of epicalyxin F was prepared and shown to be incorrect. Several calyxin natural products, including
D Raederstorff et al.
European journal of biochemistry, 164(2), 427-434 (1987-04-15)
The sterols and sterol precursors of two amoebae of the genus Naegleria, Naegleria lovaniensis and Naegleria gruberi were investigated. Cycloartenol, the sterol precursor in photosynthetic organisms, is present in both amoebae. In N. lovaniesis, it is accompanied by lanosterol and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service