Skip to Content
Merck
All Photos(1)

Key Documents

29648

Supelco

Cyclooctene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14
CAS Number:
Molecular Weight:
110.20
Beilstein:
1901031
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.470

bp

32-34 °C/12 mmHg (lit.)

density

0.848 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

neat

SMILES string

C1CCCC=CCC1

InChI

1S/C8H14/c1-2-4-6-8-7-5-3-1/h1-2H,3-8H2/b2-1-

InChI key

URYYVOIYTNXXBN-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Zhou et al.
Chemical science, 9(9), 2575-2580 (2018-05-03)
Pickering emulsion is emerging as an advanced platform for catalysis because of the large oil/water interface area for reaction and its superior efficiency. How to enhance the mass transportation within the micro-droplets is the biggest obstacle in further improving the
Chris E Freye et al.
Talanta, 211, 120668-120668 (2020-02-20)
The ability to discover minute differences between samples or sample classes for gas chromatography coupled to mass spectrometry (GC-MS) can be a challenging endeavor, especially when those differences are not a priori. Fisher ratio (F-ratio) analysis is an apt technique
Mohamed M A Soliman et al.
Molecules (Basel, Switzerland), 25(12) (2020-07-02)
Pd(II) and Pt(II) composites with activated carbon (AC), graphene oxide, and multiwalled carbon nanotubes were prepared by ball milling and used as catalysts for the Suzuki-Miyaura reaction, under several energy inputs (mechanical grinding, conventional heating, and microwave irradiation). The catalytic
Alla Zelenyuk et al.
Faraday discussions, 200, 143-164 (2017-06-06)
When secondary organic aerosol (SOA) particles are formed by ozonolysis in the presence of gas-phase polycyclic aromatic hydrocarbons (PAHs), their formation and properties are significantly different from SOA particles formed without PAHs. For all SOA precursors and all PAHs, discussed
Vasiliy Yu Evtushok et al.
Frontiers in chemistry, 7, 858-858 (2020-01-11)
In this work, we elaborated heterogeneous catalysts on the basis of the Venturello complex [PO4{WO(O2)2}4]3- (PW4) and nitrogen-free or nitrogen-doped carbon nanotubes (CNTs or N-CNTs) for epoxidation of alkenes and sulfoxidation of thioethers with aqueous hydrogen peroxide. Catalysts PW4/CNTs and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service