Skip to Content
Merck
All Photos(1)

Key Documents

04151

Supelco

Diethyl malonate

analytical standard

Synonym(s):

Malonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOC2H5)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
774687
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

5.52 (vs air)

vapor pressure

1 mmHg ( 40 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

refractive index

n20/D 1.412-1.416
n20/D 1.413 (lit.)

bp

199 °C (lit.)

mp

−51-−50 °C (lit.)

density

1.055 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCOC(=O)CC(=O)OCC

InChI

1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3

InChI key

IYXGSMUGOJNHAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Timothy Anstine et al.
Monatshefte fur chemie, 145(7), 1117-1135 (2014-08-12)
Three new linear pentapyrrole rubinoid analogs: 2,3,7,8,17,18,22,23-octamethyl-12,13-bis-(2-carboxyethyl)-1,10,15,24,25,27,28,29-octahydro-27H-pentapyrrin-1,24-dione and 2,3,8,12,13,17,22,23-octamethyl-7,18-bis-(2-carboxyethyl)-1,10,15,24,25,26,27,28,29-octahydro-27H-pentapyrrin-1,24-dione, and its 7,18-dihexanoic acid analog were synthesized, respectively, from 2,3,7,8-tetramethyl-(10H)-dipyrrin-2-one, from 2,3,8-trimethyl-7-[2-(methoxycarbonyl)ethyl]-(10H)-dipyrrinone, and 2,3,8-trimethyl-7-[5-(methoxycarbonyl)pentyl]-(10H)-dipyrrinone. 13C NMR and 1H NMR measurements in (CD3)2SO confirmed the pentapyrrole structures, while 1H NMR data indicate
Shigeo Hayashi et al.
Journal of enzyme inhibition and medicinal chemistry, 29(6), 846-867 (2014-02-13)
Because of the pivotal role of cyclooxygenase (COX) in the inflammatory processes, non-steroidal anti-inflammatory drugs (NSAIDs) that suppress COX activities have been used clinically for the treatment of inflammatory diseases/syndromes; however, traditional NSAIDs exhibit serious side-effects such as gastrointestinal damage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service