Skip to Content
Merck
All Photos(1)

Key Documents

02583

Supelco

Iron standard for AAS

analytical standard, 10.00 g/L Fe in nitric acid, traceable to BAM

Synonym(s):

Iron(III) nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10.00 g/L Fe in nitric acid

technique(s)

AAS: suitable

density

1.05 g/mL at 20 °C

format

single component solution

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Iron standard may be used as reference standard in flame atomic absorption spectrometry (FAAS) for the determination of iron in dietetic pharmaceutical preparation.[1]

Preparation Note

prepared with Fe(NO3)3.9H2O and HNO3 (~0.5 M)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of iron and molybdenum in a dietetic preparation by flame AAS after dry ashing.
Canfranc E
Journal of Pharmaceutical and Biomedical Analysis, 25(1), 103-108 (2001)
Youshan Sun et al.
Applied biochemistry and biotechnology, 164(6), 918-928 (2011-02-15)
Corn stover silage is an attractive raw material for the production of biofuels and chemicals due to its high content of carbohydrates and easy degradability. The effects of Fe(NO(3))(3) pretreatment conditions on sugar yields were investigated for corn stover silage.
Mara Ghiazza et al.
Chemical research in toxicology, 24(1), 99-110 (2010-12-22)
The mechanism of enhancement/inhibition of quartz toxicity induced by iron is still unclear. Here the amount of iron on a fibrogenic quartz (Qz) was increased by wet impregnation (Fe(NO(3))(3) 0.67 and 6.7 wt %). X-ray diffraction (XRD), XRF diffuse reflectance
Jeanet Conradie et al.
Journal of inorganic biochemistry, 100(12), 2069-2073 (2006-11-04)
All low-spin S=1/2 heme-NO complexes feature FeNO angles of about 140 degrees . In contrast, the square-pyramidal [Fe(CN)(4)(NO)](2-) complex features an exactly linear {FeNO}(7) unit. We have sought here to determine a possible, simple molecular orbital (MO) rationale for these
F Ndong et al.
International endodontic journal, 45(6), 557-564 (2012-01-25)
To determine the effect of increasing the proportion of zinc on partially stabilized cement (PSC) produced using a one-step sol gel process. A one-step sol-gel process of Portland cement-based PSC with Zn was synthesized by replacing iron nitrate. The crystalline

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service